Thermo Scientific™
8mm Autosampler Inserts
Catalog number: 02-MTV
Thermo Scientific™
8mm Autosampler Inserts
Catalog number: 02-MTV
These clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes ranging from 125-200μL.
Catalog Number
02-MTV
Unit Size
Each
Material
Glass
Volume (Metric)
200 μL
Price (USD)
Availability
***
Full specifications
Quantity
-
Description200µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
Insert TypeConical
Volume (Metric) Residual<4 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)30 mm
Quantity1000
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Marking SpotNo
Volume (Metric) Usable160 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizeEach
Catalog Number
Unit Size
Pack of 500
Material
Glass
Volume (Metric)
250 μL
Price (USD)
73.00
Your Price:
Availability
***
Full specifications
Quantity
Description200µL MicroSert Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)250 μL
FeaturesMicroSert, Flat bottom, Glass
Insert TypeFlat Bottom
Volume (Metric) Residual<12 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total250 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeMicro Insert
Volume (Metric) Usable200 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 500
Catalog Number
Unit Size
Pack of 100
Material
Glass
Volume (Metric)
200 μL
Price (USD)
80.00
Your Price:
Availability
***
Full specifications
Quantity
Description150µL Polyspring Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
FeaturesPolyspring, Conical, Pulled point, Silanized™ glass
Insert TypePulled Point
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total150 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Material
Polypropylene
Volume (Metric)
175 μL
Price (USD)
56.00
Your Price:
Availability
***
Full specifications
Quantity
Description125µL Polyspring Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialPolypropylene
Volume (Metric)175 μL
FeaturesPolyspring, Precision point, Polypropylene
Insert TypePrecision Point
Volume (Metric) Residual<2 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total175 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable125 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Material
Glass
Volume (Metric)
200 μL
Price (USD)
50.00
Your Price:
Availability
***
Full specifications
Quantity
Description150µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
FeaturesConical Insert
Insert TypePulled Point
Volume (Metric) Residual<3 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 100
Material
Glass
Volume (Metric)
200 μL
Price (USD)
113.00
Your Price:
Availability
***
Full specifications
Quantity
Description150µL Polyspring Insert, silanized
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
FeaturesPolyspring, Conical, Pulled point, Silanized™ glass
Insert TypePulled Point
Volume (Metric) Residual175 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
Catalog Number
Unit Size
Pack of 500
Material
Polyethylene
Volume (Metric)
125 µL to 200 µL
Price (USD)
63.00
Your Price:
Availability
***
Full specifications
Quantity
DescriptionSelf-centering support device for tapered glass inserts
ColorClear
Diameter (Metric) Outer8 mm
MaterialPolyethylene
Volume (Metric)125 µL to 200 µL
Insert TypeSelf-Centering Support Device
Product LineAutosampler Vial Insert
Volume (Metric) Total125 µL to 200 µL
Vial TypeInsert
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSelf-Centering Support Device
For Use With (Equipment)Tapered Glass Insert
Unit SizePack of 500
Catalog Number
Unit Size
Each
Material
Glass
Volume (Metric)
250 μL
Price (USD)
Availability
***
Full specifications
Quantity
-
Description200µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)250 μL
Insert TypeFlat Bottom
Volume (Metric) Residual<12 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity1000
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Marking SpotNo
Volume (Metric) Usable200 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizeEach
Showing 8 of 8
Catalog NumberSpecificationsUnit SizeMaterialVolume (Metric)Price (USD)AvailabilityQuantity
02-MTVFull specifications
EachGlass200 μLRequest A Quote***-
Description200µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
Insert TypeConical
Volume (Metric) Residual<4 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)30 mm
Quantity1000
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Marking SpotNo
Volume (Metric) Usable160 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizeEach
C4012-465Full specifications
Pack of 500Glass250 μL
73.00
Your Price:
***
Description200µL MicroSert Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)250 μL
FeaturesMicroSert, Flat bottom, Glass
Insert TypeFlat Bottom
Volume (Metric) Residual<12 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total250 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeMicro Insert
Volume (Metric) Usable200 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 500
C4012-530Full specifications
Pack of 100Glass200 μL
80.00
Your Price:
***
Description150µL Polyspring Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
FeaturesPolyspring, Conical, Pulled point, Silanized™ glass
Insert TypePulled Point
Volume (Metric) Residual<1 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total150 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
C4012-530PFull specifications
Pack of 100Polypropylene175 μL
56.00
Your Price:
***
Description125µL Polyspring Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialPolypropylene
Volume (Metric)175 μL
FeaturesPolyspring, Precision point, Polypropylene
Insert TypePrecision Point
Volume (Metric) Residual<2 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total175 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable125 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
C4012-529Full specifications
Pack of 100Glass200 μL
50.00
Your Price:
***
Description150µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
FeaturesConical Insert
Insert TypePulled Point
Volume (Metric) Residual<3 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
C4012-S530Full specifications
Pack of 100Glass200 μL
113.00
Your Price:
***
Description150µL Polyspring Insert, silanized
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)200 μL
FeaturesPolyspring, Conical, Pulled point, Silanized™ glass
Insert TypePulled Point
Volume (Metric) Residual175 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)29 mm
Quantity100
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Volume (Metric) Usable175 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizePack of 100
MTS-1Full specifications
Pack of 500Polyethylene125 µL to 200 µL
63.00
Your Price:
***
DescriptionSelf-centering support device for tapered glass inserts
ColorClear
Diameter (Metric) Outer8 mm
MaterialPolyethylene
Volume (Metric)125 µL to 200 µL
Insert TypeSelf-Centering Support Device
Product LineAutosampler Vial Insert
Volume (Metric) Total125 µL to 200 µL
Vial TypeInsert
Quantity500
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSelf-Centering Support Device
For Use With (Equipment)Tapered Glass Insert
Unit SizePack of 500
02-NVFull specifications
EachGlass250 μLRequest A Quote***-
Description200µL Insert
Diameter (Metric) Inner5 mm
ColorClear
Diameter (Metric) Outer5 mm
MaterialGlass
Volume (Metric)250 μL
Insert TypeFlat Bottom
Volume (Metric) Residual<12 μL
Product LineAutosampler Vial Insert
Volume (Metric) Total200 µL
Vial TypeInsert
Height (Metric)31 mm
Quantity1000
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeAutosampler Insert
Marking SpotNo
Volume (Metric) Usable200 μL
For Use With (Equipment)8 mm HPLC, GC Autosampler Vial
Unit SizeEach
Showing 8 of 8
  • Pulled point inserts are an economical choice for non-critical applications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.