Specifications
CAS34767-44-7
Chemical Name or MaterialCyclopentadienylzirconium trichloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight262.66
Health Hazard 1H302+H312+H332-H314-H500
View more