2,6-Dimethylphenyl isocyanide
2,6-Dimethylphenyl isocyanide
2,6-Dimethylphenyl isocyanide
2,6-Dimethylphenyl isocyanide
Thermo Scientific Chemicals

2,6-Dimethylphenyl isocyanide

CAS: 2769-71-3 | C9H9N | 131.18 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 020748.03
亦稱為 020748-03
價格 (TWD)
-
申請報價
Quantity:
1 g
Request bulk or custom format
規格
CAS2769-71-3
Chemical Name or Material2,6-Dimethylphenyl isocyanide
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECS Number220-459-2
Formula Weight131.18
檢視更多
2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only