Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers
Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers
Thermo Scientific™

Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers

Prevent bio-hazardous spills with these leak-resistant wide-mouth containers, ideally suited for urine collection from women during pregnancy and for older or overweight patients.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityClosure ColorLabelNo. per Case
021090
also known as 02 1090
SterileOrangeTabbed400/Cs., Pre-assembled
020180
also known as 02 0180
Non-sterileWhiteNo Label400/Cs., Bagged Separately
021016
also known as 02 1016
SterileWhiteTabbed Temp Strip300/Cs., Pre-assembled
021038
also known as 02 1038
SterileNaturalTabbed300/Cs., Pre-assembled
021051
also known as 02 1051
Non-sterileWhiteNon-tabbed300/Cs., Assembled with Tablet
021102
also known as 02 1102
Non-sterileWhiteNon-tabbed300/Cs., Pre-assembled
021104
also known as 02 1104
Non-sterileNaturalNon-tabbed300/Cs., Pre-assembled
021105
also known as 02 1105
Non-sterileYellowNon-tabbed300/Cs., Pre-assembled
021106
also known as 02 1106
Non-sterileWhiteTabbed Temp Strip300/Cs., Pre-assembled
021140
also known as 02 1140
Non-sterileNaturalNo Label300/Cs., Pre-assembled
021181
also known as 02 1181
Non-sterileWhiteNo Label300/Cs., Bagged Separately
Catalog number 021090
also known as 02 1090
Price (MXN)
-
Sterility:
Sterile
Closure Color:
Orange
Label:
Tabbed
No. per Case:
400/Cs., Pre-assembled

Prevent bio-hazardous spills with the proprietary, all plastic, leak-resistant seal of Thermo Scientific™ Samco™ Wide-Mouth Bio-Tite™ 90mL (3 oz.) 53mm Specimen Containers. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:

  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • 90mL (3 oz.); Opening: 53mm
  • Dual-threaded caps create improved sealing
  • Sterilized by gamma irradiation, which eliminates potential contamination
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Stackable containers are easy to store, saving lab space
  • Virgin polypropylene containers allow higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage
Specifications
Capacity (Metric)90 mL
Closure ColorOrange
Closure Size53 mm
Closure TypeBio-Tite Lid
DescriptionCap and container assembled
Diameter (Metric)53 mm
LabelTabbed
No. per Case400/Cs., Pre-assembled
Closure IncludedYes
MaterialVirgin Polypropylene
Product TypeSpecimen Container
ShapeCylindrical
SterilitySterile
Unit SizeCase of 400