n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl
n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl
n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl
n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl
Thermo Scientific Chemicals

n-Butyl phosphate, mixture of mono-n-butyl and di-n-butyl

CAS: 52933-01-4 | C4H11O4P | 154.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 022404.A1
also known as 022404-A1
Price (TWD)
-
Request A Quote
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS52933-01-4
IUPAC Namebutoxyphosphonic acid; dibutoxyphosphinic acid
Molecular FormulaC12H30O8P2
InChI KeyORMYKVYVDRWAKI-UHFFFAOYSA-N
SMILESCCCCOP(O)(O)=O.CCCCOP(O)(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Acid Value≥430 mg KOH/g
Specific Gravity1.115 - 1.125 (25°C)
Appearance (Color)Colorless to yellow
Color (APHA)≤100
FormLiquid
n-Butyl phosphate is a solvent and plasticizer for cellulose esters such as nitrocellulose and cellulose acetate. It forms stable hydrophobic complexes with some metals, as an extractant. It is also used in industry are as a component of aircraft hydraulic fluid and as a solvent for extraction and purification of rare earth metals from their ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only