Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid zinc disodium salt hydrate, Thermo Scientific Chemicals

Catalog number: 022426.36
500 g, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid zinc disodium salt hydrate, Thermo Scientific Chemicals

Catalog number: 022426.36
500 g, Each
Quantity
Catalog number: 022426.36
also known as 022426-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14025-21-9
IUPAC Name
zinc(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Molecular Formula
C10H12N2Na2O8Zn
InChI Key
UBLOJEHIINPTTG-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Zn++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Appearance (Color)
White to pale yellow
Assay from Suppliers CofA
≥95% to ≤105.0% (Dry wt. basis)
Loss on Drying
8.0-15.0%
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text