Thermo Scientific Chemicals

Methanol, ultrapure, HPLC Grade, 99.8+%, Thermo Scientific Chemicals

1 L, Each
Thermo Scientific Chemicals

Methanol, ultrapure, HPLC Grade, 99.8+%, Thermo Scientific Chemicals

1 L, Each
Quantity
Catalog number: 022909.K2
also known as 022909-K2
Price (USD)
Price: 43.80
Your Price:
Quantity
-

Chemical Identifiers

CAS
67-56-1
IUPAC Name
methanol
Molecular Formula
CH4O
InChI Key
OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES
CO
Comment
ACS liquid chromatography suitability: conforms [UK]
Comment
Material manufactured/sourced in either US or UK
Comment
Pesticide Residue Analysis test: pass [UK]
Comment
Sub micron filtered [US]
Optical Absorbance at
225nm: ≥80% transmittance [UK]

Description

Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin. It is also used as a mobile phase in High Performance Liquid Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only