Cyclohexane, HPLC Grade, 99% min
Cyclohexane, HPLC Grade, 99% min
Cyclohexane, HPLC Grade, 99% min
Thermo Scientific Chemicals

Cyclohexane, HPLC Grade, 99% min

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 022919.M1
also known as 022919-M1
Price (TWD)
-
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at254 nm: ≤0.01 AU
Residue on Evaporation≤1ppm
FormLiquid
Color (APHA)≤10
Assay (GC)≥99.0%
View more
Cyclohexane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only