Thermo Scientific Chemicals

Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals

Catalog number: 022922.M1
4 x 1 L, Each
Thermo Scientific Chemicals

Chlorobenzene, HPLC Grade, 99.5%, Thermo Scientific Chemicals

Catalog number: 022922.M1
4 x 1 L, Each
Quantity
Catalog number: 022922.M1
also known as 022922-M1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-90-7
IUPAC Name
chlorobenzene
Molecular Formula
C6H5Cl
InChI Key
MVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILES
ClC1=CC=CC=C1
Residual water
0.03%
Residue on Evaporation
2ppm

Description

Chlorobenzene is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text