Thermo Scientific Chemicals

Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals

1 L, Each
Thermo Scientific Chemicals

Acetonitrile, HPLC Grade, 99.7+% min, Thermo Scientific Chemicals

1 L, Each
Quantity
Catalog number: 022927.K2
also known as 022927-K2
Price (USD)
Price: 124.00
Your Price:
Quantity
-

Chemical Identifiers

CAS
75-05-8
IUPAC Name
acetonitrile
Molecular Formula
C2H3N
InChI Key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES
CC#N
Optical Absorbance at
190 nm: ≤1.00 AU
Optical Absorbance at
220 nm: ≤0.05 AU
Optical Absorbance at
235 nm: ≤0.01 AU
Form
Liquid
Assay (special)
GC-ECD Responsive Residue (single peak as heptachlorepoxide): ≤5 ppt

Description

Acetonitrile is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only