Thermo Scientific™

Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific™

4 x 4L, Each
Thermo Scientific™

Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Thermo Scientific™

4 x 4L, Each
Quantity
Unit Size
Catalog number: 022931.M6
also known as 022931-M6
Price (USD)

Chemical Identifiers

CAS
64-17-5
Color (APHA)
10, max
Assay (GC)
4 to 6% Methanol
Form
Liquid
Assay (GC)
88 to 92% Ethanol
Optical Absorbance at
210nm: 1.00AU, max

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
For Manufacturing and Laboratory Use. Not for beverage use

Figures