Tetraethoxysilane, 99.999+% (metals basis)
Tetraethoxysilane, 99.999+% (metals basis)
Tetraethoxysilane, 99.999+% (metals basis)
Thermo Scientific Chemicals

Tetraethoxysilane, 99.999+% (metals basis)

CAS: 78-10-4 | C8H20O4Si | 208.329 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 022967.30
also known as 022967-30
Price (TWD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS78-10-4
IUPAC Nametetraethyl silicate
Molecular FormulaC8H20O4Si
InChI KeyBOTDANWDWHJENH-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.001%
Tetraethoxysilane is the main precursor material for the synthesis of zeolites and silicon dioxide, which is used in semiconductor industry. It is widely used as a cross linking agent in silicon polymers and in aerogel preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only