Sulfite, Quant™ Test Strips - FAQs
View additional product information for Sulfite, Quant™ Test Strips - FAQs (023288.QA)
3 product FAQs found
What is the range of sulfite ion concentration that can be measured with the Sulfite, Quant Test Strips?
The Sulfite, Quant Test Strips can measure sulfite ion in the range of 10 to 500 ppm.
How many test strips come in the Sulfite, Quant Test Strips pack?
You will receive 100 test strips in each pack of Sulfite, Quant Test Strips.
What are the Sulfite, Quant Test Strips used for?
The Sulfite, Quant Test Strip is a semi-quantitative test for measuring sulfite (SO32-) ion in aqueous solution.