Thermo Scientific Chemicals

Copper(II) tetrafluoroborate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 026127.36
500 g, Each
Thermo Scientific Chemicals

Copper(II) tetrafluoroborate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 026127.36
500 g, Each
Quantity
Catalog number: 026127.36
also known as 026127-36
Price (USD)
Price: 331.00
Online price: 286.65
Your price:
Quantity
-

Chemical Identifiers

CAS
72259-10-0
IUPAC Name
copper(2+) bis(tetrafluoroboranuide) hexahydrate
Molecular Formula
B2CuF8H12O6
InChI Key
FBYIXFBIARQMJJ-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F
Assay (unspecified)
97.6% min

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text