Orthophosphoric acid, ≥85.0-88.0%, NF, FCC, Food GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, ≥85.0-88.0%, NF, FCC, Food GMP, J.T.Baker™

This product is GMP Manufactured and meets FCC and NF specifications.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 mL
2.5 L
4 L
70 lb.
200 lb.
700 lb.
Verpackung:
Alle anzeigen
Glasflasche
Katalognummer 0262-03
Preis (EUR)
-
Menge:
2.5 L
Verpackung:
Glasflasche
This product is GMP Manufactured and meets FCC and NF specifications.
This product is GMP Manufactured and meets FCC and NF specifications.
Specifications
CAS7664-38-2
Chemischer Name oder MaterialOrthophosphoric acid
FarbeFarblos
Schmelzpunkt28°C
SummenformelH3PO4
VerpackungGlasflasche
SynonymPhosphoric acid
GüteNF, FCC
Molekulargewicht (g/mol)98
Reinheit (%)85 to 88%
Menge2.5 L
Unit SizeEach