Orthophosphoric acid, ≥85.0-88.0%, NF, FCC, Food GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, ≥85.0-88.0%, NF, FCC, Food GMP, J.T.Baker™

This product is GMP Manufactured and meets FCC and NF specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 mL
2.5 L
4 L
70 lb.
200 lb.
700 lb.
Envase:
Ver todo
Polyethylene drum
Número de catálogo 0262-07
Precio (MXN)
-
Cantidad:
200 lb.
Envase:
Polyethylene drum
This product is GMP Manufactured and meets FCC and NF specifications.
This product is GMP Manufactured and meets FCC and NF specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
CAS7664-38-2
Nombre del producto químico o materialOrthophosphoric acid
ColorColorless
Punto de fusión28°C
Fórmula molecularH3PO4
EnvasePolyethylene drum
SinónimoPhosphoric acid
GradoNF, FCC
Peso molecular (g/mol)98
Porcentaje de pureza85 to 88%
Cantidad200 lb.
Unit SizeEach