Especificações
CAS7664-38-2
Chemical Name or MaterialOrthophosphoric acid
ColorColorless
Melting Point28°C
Molecular FormulaH3PO4
PackagingPolyethylene drum
SynonymPhosphoric acid
GradeNF, FCC
Molecular Weight (g/mol)98
Percent Purity85 to 88%
Quantity700 lb.
Unit SizeEach