Orthophosphoric acid, ≥85.0-88.0%, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Orthophosphoric acid, ≥85.0-88.0%, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets EP, BP, and NF specifications.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Quantity:
500 mL
2.5 L
4 L
700 lb.
Packaging:
Ver tudo
Clear glass bottle
Clear glass bottle, single shipper
Número do catálogo 0268-03
Preço (BRL)
-
Quantity:
2.5 L
Packaging:
Clear glass bottle
This product is GMP Manufactured and meets EP, BP, and NF specifications.
This product is GMP Manufactured and meets EP, BP, and NF specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificações
CAS7664-38-2
Chemical Name or MaterialOrthophosphoric acid
ColorColorless
Melting Point28°C
Molecular FormulaH3PO4
PackagingClear glass bottle
SynonymPhosphoric acid
GradeMulti-Compendial
Molecular Weight (g/mol)98
Percent Purity85 to 88%
Quantity2.5 L
Unit SizeEach