Colored Screw Caps
Fisher Scientific™

Colored Screw Caps

For use with nonsterile skirted- and conical-bottom tubes.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Type
02681359BlueScrew Cap with O-ring
02681358NaturalScrew Cap with O-ring
02681360YellowScrew Cap with O-ring
02681361RedScrew Cap with O-ring
02681363GreenScrew Cap with O-ring
02681364OrangeScrew Cap with O-ring
02681366VioletScrew Cap with O-ring
02681367AmberScrew Cap with O-ring
02681368
also known as FISHERMIS-02681368-EACH
AssortedScrew Cap with O-ring
Catalog number 02681359
Price (KRW)
-
Color:
Blue
Product Type:
Screw Cap with O-ring
For use with nonsterile skirted- and conical-bottom tubes
  • Feature a frosted writing surface
  • Can be pierced for sample removal
Note: For tubes, see Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tube without Caps.
Specifications
For Use With (Equipment)Non-sterile skirted and conical bottom tubes
IncludesO-ring
AutoclavableAutoclavable
ColorBlue
MaterialPolypropylene
Product TypeScrew Cap with O-ring
SterilityNon-sterile
Unit SizePack of 500