Europium(III) sulfate octahydrate, 99.9% (REO)
Europium(III) sulfate octahydrate, 99.9% (REO)
Europium(III) sulfate octahydrate, 99.9% (REO)
Thermo Scientific Chemicals

Europium(III) sulfate octahydrate, 99.9% (REO)

CAS: 10031-55-7 | Eu2H16O20S3 | 736.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 029106.04
also known as 029106-04
Price (TWD)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS10031-55-7
IUPAC Namedieuropium(3+) octahydrate trisulfate
Molecular FormulaEu2H16O20S3
InChI KeyPGDXJFDXKJLVTJ-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Eu+3].[Eu+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Europium(III) sulfate octahydrate is used as a phosphor activator for color cathode-ray tubes and liquid-crystal displays. It is used in computer monitors and televisions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only