Certified Cleaned Straight Sided Bottles
Certified Cleaned Straight Sided Bottles
Certified Cleaned Straight Sided Bottles
Certified Cleaned Straight Sided Bottles
Fisher Scientific™
Certified Cleaned Straight Sided Bottles
Ideal for soil sampling and environmental applications. Fisherbrand™ Certified Cleaned Straight Sided Bottles are manufactured from clear soda glass.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)
02912312500 mL
02912310125 mL
029123131 L
카탈로그 번호 02912312
제품 가격(KRW)
-
용량(미터법):
500 mL
Ideal for soil sampling and environmental applications. Fisherbrand™ Certified Cleaned Straight Sided Bottles are manufactured from clear soda glass.
  • Cleaned per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
  • Straight sided walls allow for the complete removal of contents
  • Bottles are autoclavable; caps are not
사양
마개 색상White
마개 크기89-400 mm
마개 유형Screw Cap
길이(미터법)55mm
라이너 재질PTFE
수량12/PK.
용량(영국식 단위)16 oz.
용량(미터법)500 mL
인증/규정준수Certificate of analysis
Closure IncludedYes
색상Clear
물질Soda Lime Glass
Unit SizeCase of 12