02GOLD also known as 02 GOLD
Full specifications
Each Vial Request A Quote - - Diameter (English) Inner 0.9 in. Diameter (Metric) Inner 23 mm Color Gold Material Polypropylene Sterility Sterile Volume (Metric) 60 mL Height (English) 3.3 in. Volume (Metric) Total 60 mL Closure Type Flip Top Height (Metric) 84 mm Quantity 1 Ea. Certifications/Compliance FDA Compliant Type Vial Unit Size Each
0385 Full specifications
Case of 400 90mL (3 oz.); With skirt top Skirt top Polypropylene Container Request A Quote - - Description 90mL (3 oz.); With skirt top Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Skirt top Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Type Polypropylene Container Unit Size Case of 400
04HP105B-WHITE Full specifications
Case of 240 4 oz. (118 mL); High Profile; White High Profile Polypropylene Container - Description 4 oz. (118 mL); High Profile; White Diameter (Metric) Inner 43 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style High Profile Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 90 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 240
03CLFREGORED Full specifications
Case of 400 3 oz. (90mL); Frego Red Standard Polypropylene Container Request A Quote - - Description 3 oz. (90mL); Frego Red Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Standard Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 400
03CLGOLD Full specifications
Each 3 oz. (90mL); Gold Standard Polypropylene Container Request A Quote - - Description 3 oz. (90mL); Gold Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Standard Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Each
03EDM33 Full specifications
Case of 400 90mL (3 oz.); With write-on space Write-on space Polypropylene Container - Description 90mL (3 oz.); With write-on space Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Write-on space Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Type Polypropylene Container Unit Size Case of 400
04HPLTRED also known as 04HP LTRED
Full specifications
Each 4 oz. (118mL); High profile; Light red High profile Vial Request A Quote - - Description 4 oz. (118mL); High profile; Light red Color Light Red Material Polypropylene Sterility Aseptic Volume (Metric) 118 mL Style High profile Volume (English) 4 oz. Volume (Metric) Total 118 mL Closure Type Flip Top Quantity 1 Ea. Certifications/Compliance FDA Compliant Type Vial Unit Size Each
04HPWHITE also known as 04HP WHITE
Full specifications
Each 4 oz. (118mL); High profile; White High profile Vial Request A Quote - - Description 4 oz. (118mL); High profile; White Color White Material Polypropylene Sterility Aseptic Volume (Metric) 118 mL Level FDA approved Style High profile Volume (English) 4 oz. Volume (Metric) Total 118 mL Closure Type Flip Top Quantity 1 Ea. Certifications/Compliance FDA Compliant Type Vial Unit Size Each
04HP-SMOOTH Full specifications
Case of 250 4 oz. (118mL); High profile; With write-on space High profile with write-on space Polypropylene Container Contact Us › - - Description 4 oz. (118mL); High profile; With write-on space Diameter (Metric) Inner 43 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style High profile with write-on space Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 90 mm Type Polypropylene Container Unit Size Case of 250
04LP Full specifications
Case of 250 4 oz. (118mL); Low profile; Natural Low profile Polypropylene Container - Description 4 oz. (118mL); Low profile; Natural Diameter (Metric) Inner 63 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style Low profile Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 50 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 250