02LTBROWN also known as 02 LTBROWN
Full specifications
Each Vial Request A Quote - - Diameter (English) Inner 0.9 in. Diameter (Metric) Inner 23 mm Color Light Brown Material Polypropylene Sterility Sterile Height (English) 3.3 in. Closure Type Flip Top Height (Metric) 84 mm Quantity 1 Ea. Certifications/Compliance FDA Compliant Type Vial Unit Size Each
0385 Full specifications
Case of 400 90mL (3 oz.); With skirt top Skirt top Polypropylene Container Request A Quote - - Description 90mL (3 oz.); With skirt top Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Skirt top Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Type Polypropylene Container Unit Size Case of 400
04HP105B-WHITE Full specifications
Case of 240 4 oz. (118 mL); High Profile; White High Profile Polypropylene Container - Description 4 oz. (118 mL); High Profile; White Diameter (Metric) Inner 43 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style High Profile Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 90 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 240
03CLFREGORED Full specifications
Case of 400 3 oz. (90mL); Frego Red Standard Polypropylene Container Request A Quote - - Description 3 oz. (90mL); Frego Red Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Standard Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 400
03CLGOLD Full specifications
Each 3 oz. (90mL); Gold Standard Polypropylene Container Request A Quote - - Description 3 oz. (90mL); Gold Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Standard Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Each
03EDM33 Full specifications
Case of 400 90mL (3 oz.); With write-on space Write-on space Polypropylene Container - Description 90mL (3 oz.); With write-on space Diameter (Metric) Inner 33 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 90 mL No. per Case 400 Style Write-on space Volume (English) 3 oz. Closure Type Flip Top Height (Metric) 95 mm Type Polypropylene Container Unit Size Case of 400
04HPLTRED also known as 04HP LTRED
Full specifications
Each 4 oz. (118mL); High profile; Light red High profile Vial Request A Quote - - Description 4 oz. (118mL); High profile; Light red Color Light Red Material Polypropylene Sterility Aseptic Volume (Metric) 118 mL Style High profile Volume (English) 4 oz. Volume (Metric) Total 118 mL Closure Type Flip Top Quantity 1 Ea. Certifications/Compliance FDA Compliant Type Vial Unit Size Each
04HPWHITE also known as 04HP WHITE
Full specifications
Each 4 oz. (118mL); High profile; White High profile Vial Request A Quote - - Description 4 oz. (118mL); High profile; White Color White Material Polypropylene Sterility Aseptic Volume (Metric) 118 mL Level FDA approved Style High profile Volume (English) 4 oz. Volume (Metric) Total 118 mL Closure Type Flip Top Quantity 1 Ea. Certifications/Compliance FDA Compliant Type Vial Unit Size Each
04HP-SMOOTH Full specifications
Case of 250 4 oz. (118mL); High profile; With write-on space High profile with write-on space Polypropylene Container Contact Us › - - Description 4 oz. (118mL); High profile; With write-on space Diameter (Metric) Inner 43 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style High profile with write-on space Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 90 mm Type Polypropylene Container Unit Size Case of 250
04LP Full specifications
Case of 250 4 oz. (118mL); Low profile; Natural Low profile Polypropylene Container - Description 4 oz. (118mL); Low profile; Natural Diameter (Metric) Inner 63 mm Color Natural Material Polypropylene Sterility Non-sterile Volume (Metric) 118 mL No. per Case 250 Style Low profile Volume (English) 4 oz. Closure Type Flip Top Height (Metric) 50 mm Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC) Type Polypropylene Container Unit Size Case of 250