Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve.
Have Questions?
Catalog number 03-052-409
Price (USD)/ Each
-
Request A Quote
Use the Thermo Scientific™ Valco™ Injector Model C6W as an injector or switching valve. It is actuated manually and has 1/16 in. fittings and six 0.010 in. ports.