Search
View additional product information for Al-23 Tube (one end closed);OD (mm), 4;ID (mm), 2 - FAQs (030062.EM, 030062.DW, 030062.EH, 030062.DR, 030062.EO)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.