Search
전체검색
View additional product information for Al-23 Tube (one end closed);OD (mm), 30;ID (mm), 25 - FAQs (030080.EH)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.