Thermo Scientific Chemicals

Di-tert-butyl phosphite, 95%, Thermo Scientific Chemicals

Catalog number: 030233.06
5 g, Each
Thermo Scientific Chemicals

Di-tert-butyl phosphite, 95%, Thermo Scientific Chemicals

Catalog number: 030233.06
5 g, Each
Quantity
Catalog number: 030233.06
also known as 030233-06
Price (USD)
Price: 63.20
Online price: 54.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13086-84-5
IUPAC Name
bis(tert-butoxy)(oxo)-λ⁵-phosphanylium
Molecular Formula
C8H18O3P
InChI Key
GEBLOQXLELCEEO-UHFFFAOYSA-N
SMILES
CC(C)(C)O[P+](=O)OC(C)(C)C
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (unspecified)
> 94.0%

Description

Di-tert-butyl phosphite is used as a solvent, as an antioxidant, and as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text