Tricyclohexylphosphine, 96%
Tricyclohexylphosphine, 96%
Tricyclohexylphosphine, 96%
Thermo Scientific Chemicals

Tricyclohexylphosphine, 96%

CAS: 2622-14-2 | C18H33P | 280.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 030386.14
also known as 030386-14
Price (EUR)
365,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
365,00
Each
Specifications
CAS2622-14-2
Chemical Name or MaterialTricyclohexylphosphine
ColorWhite
EINECS Number220-069-2
Formula Weight280.42
View more
Tricyclohexylphosphine is used as a ligand in organometallic chemistry. As a reagent, it is used for a range of metal-catalyzed organic transformations. Crabtree's catalyst is an organoiridium compound containing tricyclohexyl phosphine used for hydrogenation of mono-, di-, tri-, and tetra- substituted substrates and hydrogen transfer reactions. Grubbs’catalysts consists of transition metal ruthenium and tricylohexyl phosphine utilized in olefin metathesis in synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only