Iron(II) ethylenediammonium sulfate tetrahydrate, 99%
Thermo Scientific Chemicals

Iron(II) ethylenediammonium sulfate tetrahydrate, 99%

CAS: 113193-60-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 030547.14
also known as 030547-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS113193-60-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale green
Assay (Titration)> 98.5%
Chloride content< 0.005%
FormCrystals
Iron(II) ethylenediammonium sulfate tetrahydrate is used in the preparation of gold nanoparticles by reacting with gold(III)-bis[ethylene-diamine]-chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only