Aluminum sec-butoxide, 95%
Aluminum sec-butoxide, 95%
Aluminum sec-butoxide, 95%
Thermo Scientific Chemicals

Aluminum sec-butoxide, 95%

CAS: 2269-22-9 | C12H30AlO3 | 249.351 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
4 x 500 g
Catalog number 030721.36
also known as 030721-36
Price (EUR)
79,10
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
79,10
Each
Chemical Identifiers
CAS2269-22-9
IUPAC Namealuminium(3+) tris(butan-2-olate)
Molecular FormulaC12H27AlO3
InChI KeyWOZZOSDBXABUFO-UHFFFAOYNA-N
SMILES[Al+3].CCC(C)[O-].CCC(C)[O-].CCC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormViscous liquid
Assay from Supplier's CofA≥94.0 to ≤106.0%
Appearance (Color)Clear to faintly turbid colorless to pale yellow
It is used for active catalyst and support, inorganic membrane, redrcing agent of aldehyde-ketone, oxidant of aldehyde-ketone, gel forming agent of printing ink and paint, dehydrant, water proofing agent and aluminum base grease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only