Thermo Scientific Chemicals

Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT, Thermo Scientific Chemicals

500 mL, Each
Thermo Scientific Chemicals

Tetrahydrofuran, ACS, 99+%, stab. with 250ppm BHT, Thermo Scientific Chemicals

500 mL, Each
Quantity
Catalog number: 030760.AP
also known as 030760-AP
Price (USD)
Price: 40.90
Your Price:
Quantity
-

Chemical Identifiers

CAS
109-99-9
IUPAC Name
oxolane
Molecular Formula
C4H8O
InChI Key
WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES
C1CCOC1
Appearance (Color)
Clear colorless
Form
Liquid
Color (APHA)
20, max
Assay (GC)
≥99.0%
Water Content (Karl Fischer Titration)
0.05%, max

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Maximum level of impurities: Color (APHA) 20, Peroxide (as H2O2) 0.015%, Residue after evaporation 0.03%, H2O 0.05%
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only