Tetramethylammonium hydroxide, 25% w/w in methanol
Tetramethylammonium hydroxide, 25% w/w in methanol
Tetramethylammonium hydroxide, 25% w/w in methanol
Thermo Scientific Chemicals

Tetramethylammonium hydroxide, 25% w/w in methanol

CAS: 75-59-2 | C4H13NO | 91.154 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
10 g
100 g
500 g
產品號碼 030833.36
亦稱為 030833-36
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS75-59-2
IUPAC Nametetramethylazanium hydroxide
Molecular FormulaC4H13NO
InChI KeyWGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow
Assay from Suppliers CofA24.0-26.0%
FormLiquid
Tetramethylammonium hydroxide is used for anisotropic etching of silicon. As a basic solvent, it is used in the development of acidic photoresist in the photolithography process. It acts as phase transfer catalyst and surfactant and is used in the synthesis of ferrofluid in order to inhibit nanoparticle aggregation. It finds application in thermochemolysis, an analytical technique involving both pyrolysis and chemical derivatization of the analyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only