1-Pentanol, ACS, 98+%
1-Pentanol, ACS, 98+%
1-Pentanol, ACS, 98+%
Thermo Scientific Chemicals

1-Pentanol, ACS, 98+%

Filtered through 0.2μ filters. Suitable for spectrophotometry, chromatography | CAS: 71-41-0 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
Catalog number 030898.AK
also known as 030898-AK
Price (EUR)
41,30
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
41,30
Each
Chemical Identifiers
CAS71-41-0
IUPAC Namepentan-1-ol
Molecular FormulaC5H12O
InChI KeyAMQJEAYHLZJPGS-UHFFFAOYSA-N
SMILESCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)≤30
Assay (special)≥98.0%
Appearance (Color)Clear colorless
Residual water≤0.5%
Impurity contentAcids and esters: ≤0.075meq/g
View more
1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only