Bis(dicarbonylcyclopentadienyliron), 99%
Bis(dicarbonylcyclopentadienyliron), 99%
Bis(dicarbonylcyclopentadienyliron), 99%
Thermo Scientific Chemicals

Bis(dicarbonylcyclopentadienyliron), 99%

CAS: 12154-95-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 031114.14
also known as 031114-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
CAS12154-95-9
Chemical Name or MaterialBis(dicarbonylcyclopentadienyliron)
ColorRed
DOT InformationHazard Class: 6.1; Packaging Group: III
EINECS Number235-276-3
View more
Cyclopentadienyliron dicarbonyl dimer is a workhorse in organometallic chemistry because it is inexpensive and rugged. It is also used as a pharmaceutical intermediates and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only