5-Chloro-1,10-phenanthroline
5-Chloro-1,10-phenanthroline
5-Chloro-1,10-phenanthroline
Thermo Scientific Chemicals

5-Chloro-1,10-phenanthroline

CAS: 4199-89-7 | C12H7ClN2 | 214.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 031180.02
also known as 031180-02
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.70
Save 13.05 (15%)
Each
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 30-Jun-2026
85.70
Save 13.05 (15%)
Each
Specifications
CAS4199-89-7
Chemical Name or Material5-Chloro-1,10-phenanthroline
EINECS Number224-098-1
Formula Weight214.66
IUPAC Name5-chloro-1,10-phenanthroline
View more
Used to prepare 1,10-phenanthrolinium cation. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only