Sodium hydroxide solution, 10 N, WFI, Biotech Reagent, GMP, J.T.Baker™
Avantor™

Sodium hydroxide solution, 10 N, WFI, Biotech Reagent, GMP, J.T.Baker™

This product is GMP Manufactured, made from water for injection USP and sodium hydroxide pellets, NF (Multi-Compendial). Meets BP, Ph. Eur. and JP chemical specifications.
Have Questions?
Número de catálogoCantidad
0312-0719 L
Número de catálogo 0312-07
Precio (MXN)
-
Cantidad:
19 L
This product is GMP Manufactured, made from water for injection USP and sodium hydroxide pellets, NF (Multi-Compendial). Meets BP, Ph. Eur. and JP chemical specifications.
This product is GMP Manufactured, made from water for injection USP and sodium hydroxide pellets, NF (Multi-Compendial). Meets BP, Ph. Eur. and JP chemical specifications.

Para su uso en investigación o procesos de fabricación posteriores. No apto para uso diagnóstico ni para la administración directa en seres humanos ni en animales.

Especificaciones
Punto de ebullición1429°C (1013 hPa)
CAS1310-73-2
Nombre del producto químico o materialSodium hydroxide solution
ColorColorless
Punto de fusión1429°C (1013 hPa)
Fórmula molecularNaOH
EnvasePoly pail (Jerrican)
SinónimoSodium hydrate, Soda lye, Caustic soda, Lye
GradoBiotech
Peso molecular (g/mol)40
Cantidad19 L
Unit SizeEach