Thermo Scientific Chemicals

Di-n-butyl phthalate, 99+%, Thermo Scientific Chemicals

Catalog number: 031244.A1
1 kg, Each
Thermo Scientific Chemicals

Di-n-butyl phthalate, 99+%, Thermo Scientific Chemicals

Catalog number: 031244.A1
1 kg, Each
Quantity
Catalog number: 031244.A1
also known as 031244-A1
Price (USD)

Chemical Identifiers

CAS
84-74-2
IUPAC Name
1,2-dibutyl benzene-1,2-dicarboxylate
Molecular Formula
C16H22O4
InChI Key
DOIRQSBPFJWKBE-UHFFFAOYSA-N
SMILES
CCCCOC(=O)C1=CC=CC=C1C(=O)OCCCC
Appearance (Color)
Clear colorless to pale yellow
Assay from Supplier's CofA
≥99.0%
Acid Value
≤0.2% (as phthalic acid)
Form
Liquid

Description

Dibutyl phthalate (DBP) is a common industrial chemicals used as plasticizers, softeners, adhesives or solvents used by a variety of industries. It is also used as an additive to adhesives or printing inks. Other applications include in PVC, paints, cosmetics and coatings of cars.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text