Thermo Scientific Chemicals

Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals

500 mL, Each
Thermo Scientific Chemicals

Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals

500 mL, Each
Quantity
Catalog number: 031344.AP
also known as 031344-AP
Price (USD)
Price: 33.60
Your Price:
Quantity
-

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Appearance (Color)
Colorless
Form
Liquid
Color (APHA)
≤10
Assay (GC)
≥99.5% (corrected for water)
Free acid (titration)
≤0.0009meq/g

Description

Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only