Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1.6;ID (mm), 0.8, Thermo Scientific Chemicals - FAQs
View additional product information for Al-23 Insulating Tube, Both Ends Open (Thin Wall);OD (mm), 1.6;ID (mm), 0.8 - FAQs (031485.DW, 031485.DR, 031485.QAT)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.