Al-23 Tube (both ends open);OD (mm), 7;ID (mm), 4 - FAQs
Al-23 Tube (both ends open);OD (mm), 7;ID (mm), 4 - FAQs
View additional product information for Al-23 Tube (both ends open);OD (mm), 7;ID (mm), 4 - FAQs (031558.DW, 031558.EO, 031558.DM)
1 product FAQs found
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.