Search
Buscar
View additional product information for Al-23 Tube (both ends open);OD (mm), 11;ID (mm), 7 - FAQs (031563.EM, 031563.EH, 031563.QAT)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.