n-Hexatriacontane, 97+%
n-Hexatriacontane, 97+%
n-Hexatriacontane, 97+%
Thermo Scientific Chemicals

n-Hexatriacontane, 97+%

CAS: 630-06-8 | C36H74 | 506.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 031641.09
also known as 031641-09
Price (EUR)
68,90
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
68,90
Each
Chemical Identifiers
CAS630-06-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)71.0-77.0°C
Assay (GC)≥97.0%
Appearance (Color)White to off-white
FormPowder or wax or flakes
n-Hexatriacontane is used as a paraffin wax and in candles. It is used as a key starting material for the preparation of lower alkanes by cracking. It is also used for example in road surfacing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only