Search
Buscar
View additional product information for Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 8;ID (mm), 7 - FAQs (031708.DR, 031708.DW, 031708.DM)
1 product FAQs found
The maximum working temperature for the Al-23 Tube is 1950 degrees C.