Mineral oil
Thermo Scientific Chemicals

Mineral oil

CAS: 8020-83-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 x 1 kg
Catalog number 031911.A1
also known as 031911-A1
Price (EUR)
71,65
Online exclusive
79,90
Save 8,25
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
71,65
Online exclusive
79,90
Save 8,25
Each
Chemical Identifiers
CAS8042-47-5
SpecificationsSpecification SheetSpecification Sheet
Specific Gravity0.8490 to 0.8650 ( at 25?C)
Viscosity23.90 - 27.01cSt at 40?C
Mineral oil is used as an overlay covering microdrops of culture medium in petri dishes. It is an active ingredient in baby lotions, cold creams, ointments and cosmetics. It is also used in electrical and mechanical industries as thermal fluid in electrical components and non -conductive coolant. It finds application as a transformer oil, as an anti rust agent for blades and as an insulator in high voltage switchgear. Further, it is used to store reactive metals like lithium, sodium, etc and in X-ray crystallography to suspend crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only