n-Octadecane, 99%
n-Octadecane, 99%
n-Octadecane, 99%
Thermo Scientific Chemicals

n-Octadecane, 99%

CAS: 593-45-3 | C18H38 | 254.502 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 031954.22
also known as 031954-22
Price (EUR)
52,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
52,10
Each
Chemical Identifiers
CAS593-45-3
IUPAC Nameoctadecane
Molecular FormulaC18H38
InChI KeyRZJRJXONCZWCBN-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLow melting solid
Assay from Supplier's CofA>99%
Appearance (Color)White
n-octadecane is used as solvent, lubricant, transformer oil and anti-corrosion agents. It is also used in paraffin as well as chemical intermediate in organic synthesis. It plays an important role in pheromones as a chemical messenger. N-Octadecane/ molecular sieve 5A composites find application as thermal storage material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only