1,2-Dichlorobenzene, Spectrophotometric Grade, 98%
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%
Thermo Scientific Chemicals

1,2-Dichlorobenzene, Spectrophotometric Grade, 98%

Maximum level of impurities: Evaporation residue 5ppm, H2O 0.02% | CAS: 95-50-1 | C6H4Cl2 | 146.998 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 032154.M1
also known as 032154-M1
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (USD)
299.65
Online Exclusive
333.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS95-50-1
IUPAC Name1,2-dichlorobenzene
Molecular FormulaC6H4Cl2
InChI KeyRFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water0.02%
Residue on Evaporation5ppm
1,2-Dichlorobenzene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only