Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads
Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads
Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads
Thermo Scientific Chemicals

Molecular sieves, 4A with indicator, -8+12 (ca 2mm) beads

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 032256.A1
also known as 032256-A1
Price (EUR)
160,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
160,00
Each
Chemical Identifiers
CAS70955-01-0
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
SpecificationsSpecification SheetSpecification Sheet
Moisture holding capacity20 wt% water capacity, min
Appearance (Color)Blue-Tan
Formbeads

Molecular sieves, 4 Å… is used to absorb molecules as large as butane. It is utilized for the separation of natural gas, liquefied petroleum gas (LPG) and alkenes. It finds application in the refining and purification of liquid and gases. It is also used for drying and purification of air, inert gases and solvents. It is actively involved to remove methanol, carbon dioxide and ammonia from fluid streams. It plays a vital role to remove heavy metal ions available in water. It is also used as a soap forming agent and as a detergent auxiliary. It has been considered as a general-purpose drying agent in polar and non polar media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only