N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals
N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 032312.06
also known as 032312-06
Price (USD)/ Each
350.65
Online exclusive
412.00 
Save 61.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
350.65
Online exclusive
412.00 
Save 61.35 (15%)
Add to cart
N,N-Bis(2-hydroxyethyl)ethylenediamine, Thermo Scientific Chemicals
Catalog number032312.06
Price (USD)/ Each
350.65
Online exclusive
412.00 
Save 61.35 (15%)
-
Add to cart
Specifications
Chemical Name or MaterialN,N-Bis(2-hydroxyethyl)ethylenediamine
CAS3197-06-6
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
It is employed as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only