Thermo Scientific Chemicals

Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 032436.K2
1 L, Each
Thermo Scientific Chemicals

Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Catalog number: 032436.K2
1 L, Each
Quantity
Catalog number: 032436.K2
also known as 032436-K2
Price (USD)
Price: 150.00
Online price: 127.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-86-1
IUPAC Name
pyridine
Molecular Formula
C5H5N
InChI Key
JUJWROOIHBZHMG-UHFFFAOYSA-N
SMILES
C1=CC=NC=C1
Residual water
0.07%
Residue on Evaporation
4ppm

Description

Pyridine is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text