Acetonitrile, Spectrophotometric Grade, 99.7+%
Acetonitrile, Spectrophotometric Grade, 99.7+%
Acetonitrile, Spectrophotometric Grade, 99.7+%
Thermo Scientific Chemicals

Acetonitrile, Spectrophotometric Grade, 99.7+%

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
Catalog number 032470.M1
also known as 032470-M1
Price (EUR)
441,00
Each
Quantity:
4 x 1 L
Request bulk or custom format
Price (EUR)
441,00
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Absorbance at193nm: ≥50% transmittance [UK]
Optical Absorbance at190nm: ≤1.00 AU [US] / ≥18% transmittance [UK]
Optical Absorbance at197nm: ≥80% transmittance [UK]
Optical Absorbance at210nm: ≤0.10 AU [US]
Optical Absorbance at215nm: ≥95% transmittance [UK]
View more
Acetonitrile is utilized as a polar aprotic solvent in organic synthesis, and in the purification of butadiene. It is used in all the analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS). It is used for the extraction of fatty acids, spinning fibers and casting and molding of plastic materials. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins. It acts as a stabilizer for the chlorinated solvents, and finds use in the production of DNA oligonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only